ContactUsLinkComponent
PYRUVATE OXIDASE. From: Microorganism (EC 1.2.3.3) Lyophilized powder Activity: Not less than 1.5 units/mg-solid Unit Definition: On e unit causes the formation of one micromole of hydrogen peroxide (half a micromole of quinoneimine dye) per minute 1 * 100 AU
Catalog # ICNA0215348680
Supplier:  MP Biomedicals
PYRUVATE OXIDASE. From: Microorganism (EC 1.2.3.3) Lyophilized powder Activity: Not less than 1.5 units/mg-solid Unit Definition: On e unit causes the formation of one micromole of hydrogen peroxide (half a micromole of quinoneimine dye) per minute 1 * 100 AU
Catalog # ICNA0215348680
Supplier:  MP Biomedicals
Frequently Bought Together