Bromochlorophenol blue, powder
Product Details & Documents
Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
Soluble in ethanol (0,1% w/v - clear yellow solution), methanol (40 mg/4 ml), EGME, water, benzene.
- CAS Nummer:2553-71-1
- Formel:C₁₉H₁₀Br₂Cl₂O₅S
- MDL-Nummer:MFCD00047291
- Molekulargewicht:581.06 g/mol
- Siedepunkt:603 °C (1013 hPa)
- Schmelzender Platingehalt:273 °C
- EINECS:219-861-0
Technische Daten
Technische Daten
- Pack type:Glass bottle for solids
- Pk:5 g
Technische Daten
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



