ContactUsLinkComponent
SC 150L (230V/50Hz) Immersion circulators with clamp Amb+13 to 150 C, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB, Safety class 3 (FL), 45mm longer immersion depth, 1 * 1 items
Catalog # HAKA1540011
SC 150L (230V/50Hz) Immersion circulators with clamp Amb+13 to 150 C, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB, Safety class 3 (FL), 45mm longer immersion depth, 1 * 1 items
Catalog # HAKA1540011
Frequently Bought Together