ContactUsLinkComponent
AC 150-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC150-S7, Amb+13 TO +150 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PU MP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT 8 LANGUAGES UL/CSA, 1 * 1 items
  HAKA1551071
AC 150-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC150-S7, Amb+13 TO +150 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PU MP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT 8 LANGUAGES UL/CSA, 1 * 1 items
  HAKA1551071
Frequently Bought Together