AC 150-S14P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, AC150-S14P, Amb+13 to 100 C, 8-14 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, F ORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1553141
Supplier: THERMO ELECTRON LED
Frequently Bought Together