ContactUsLinkComponent
AC 150-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC150-S13, Amb+13 TO +150 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT 8 LANGUAGES UL/CSA, 1 * 1 items
Catalog # HAKA1551131
AC 150-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC150-S13, Amb+13 TO +150 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT 8 LANGUAGES UL/CSA, 1 * 1 items
Catalog # HAKA1551131
Frequently Bought Together