ContactUsLinkComponent
AC 200-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S7, Amb+13 TO +200 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PU MP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES UL/ 1 * 1 items
Catalog # HAKA1561071
AC 200-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S7, Amb+13 TO +200 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PU MP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES UL/ 1 * 1 items
Catalog # HAKA1561071
Frequently Bought Together