ContactUsLinkComponent
AC 200-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S13, Amb+13 TO +200 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1561131
AC 200-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S13, Amb+13 TO +200 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1561131
Frequently Bought Together