ContactUsLinkComponent
AC 200-A25B (230V/50Hz) OPEN REFRIGERATION BATH CIRCULATOR STEEL, AC200-A25B, -25 to +200 DEGREE C, 500 W COOLING CAPACITY AT 20C, 13-21 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB 1 * 1 items
  HAKA1564251
AC 200-A25B (230V/50Hz) OPEN REFRIGERATION BATH CIRCULATOR STEEL, AC200-A25B, -25 to +200 DEGREE C, 500 W COOLING CAPACITY AT 20C, 13-21 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB 1 * 1 items
  HAKA1564251
Frequently Bought Together