Orders LinkContactUsLinkComponent
Software for spectrometers
Software for spectrometers
Datrys™ CFR software, Ultrospec™ 7000/8000/9000
Catalog # SCLI29003624
Supplier:   BIOCHROM
Software for spectrometers
Catalog # SCLI29003624
Supplier:  BIOCHROM
Software for spectrometers
Datrys™ CFR software, Ultrospec™ 7000/8000/9000
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
This software is supplied in different modules to meet the application requirements of different customer groups and to offer a simple upgrade path should your requirements change.
  • Datrys (standard) software for routine measurement analysis and PC control
  • Datrys Lite software for quick read, fast scanning and PC control
  • Datrys Life Science software for nucleic acids, proteins and cell density measurement analysis and PC control
  • Datrys CFR software supports 21 cfr part 11 compatibility, routine measurement analysis and PC control
For many users, having an external PC to control their spectrophotometer and manipulate data, gives them the ultimate in flexibility and control. Whether looking for small differences in multiple spectral overlays, carrying out post-run manipulations on large numbers of samples, Datrys software has the flexibility to work in the way you want.
Specifications
  • Description:
    Datrys™ CFR software
  • Seq:
    04
  • For:
    Ultrospec™ 7000/8000/9000
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..