Cables
InLab® cable S7-RCA, 1,2 m
Katalognr. 661-0266
Anbieter: Mettler - Toledo
Direkt zu
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Dokumente
Mettler InLab® Sensors
Produktliteratur
Technische Daten
- Description:InLab® cable S7-RCA
- Length:1,2 m
- Seq:167
- Cat. No.:661-0266