Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SPONGE CLOTH SMALL RED 200 X 180 MM 1 * 10 items
Catalog # KAMM2137800
Supplier:
KAMMERER
Expand image
SPONGE CLOTH SMALL RED 200 X 180 MM 1 * 10 items
Catalog # KAMM2137800
Supplier:
KAMMERER
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Accessories for dissolved oxygen probes
Supplier: WTW
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Laboratory Markers
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Latex Gloves
Supplier: VWR Collection