Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Autoclaves, top loading

Autoclave, top loading, 3870 ELV-WR-D, with water reservoir and advanced microprocessor control panel, 85 L
Catalog # 481-0905
Supplier:   TUTTNAUER
Autoclaves, top loading
Catalog # 481-0905
Supplier:  TUTTNAUER
Autoclaves, top loading
Autoclaves, top loading

Autoclave, top loading, 3870 ELV-WR-D, with water reservoir and advanced microprocessor control panel, 85 L

Price
24 600.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceutical, food, medical and biotechnological industries. The advanced laboratory line is a quality range of reliable bench top (front loading) and vertical (top loading) models with chamber volumes from 28 to 160 litres, and provides a single solution for the full spectrum of sterilisation needs. The autoclaves can be fully configured from general use to customer-specific applications. Units from either the Essential or Hi End ranges can be customised with various different optional features to meet more specific or demanding applications. The sterilising temperature range is 105 to 138 °C.

  • High quality AISI 316L stainless steel chamber with mirror finished door
  • Advanced control system with easy to use 3-button keypad
  • Up to 20 customisable cycle programs
  • Standard USB and Ethernet connection allows easy transfer of cycle data to PC
  • Two flexible PT100 temperature sensors
  • AISI 304 stainless steel housing

Essential vertical (top loading) laboratory autoclaves are floor standing models. The ELV-D models are designed to be connected to utilities, and are well known for their steam production inside the chamber by using heating element at the bottom of the chamber. Allowing the user to run basic sterilisation cycles for solid loads, liquids and waste (solid or liquid) Adding different options and accessories will allow the user to expect even more from the autoclave, such as quicker cooling times and safe handling of hazardous loads.
* Automatic filling and draining of the chamber
* Automatic cooling of the exhaust to the drain 60 °C (by parameter)
'Stand alone' ELV-WR-D models have a water reservoir and only need power connection.

Hi End vertical (top loading) laboratory autoclaves are floor standing models. The ELVC-GD models are designed to be connected to utilities, and are well known for their steam production in the built-in steam generator as well as the standard available fast cooling option. Allowing the user to run besides the basic sterilisation cycles for solid loads, liquids and waste (solid or liquid) also liquid cycles with fast cooling. Adding different options and accessories will allow the user to expect even more from the autoclave such as even quicker cooling times, dry products at the end and; safe handling of hazardous loads.
* Full utilities required (tap- and DEMI-water, drain connection, compressed air and electricity)
* Automatic filling and draining functionality
* Automatic cooling of the exhaust to the drain 60 °C (by parameter)

 : Accessories as well as a range of factory fitted options such as units with fast/air cooling, Super fast cooling, pre-/post-vacuum and models suitable for bio-hazardous waste sterilisation are available on request. Please contact your local Avantor sales office for further details prior to ordering as these cannot be retro-fitted.

 : Note: All 2840 and Essential 'stand-alone' 3850 models require 230 V single-phase and the Essential and Hi End 3850 plus all 3870, 5050 and 5075 plus Essential 'stand-alone' 3870 and 5075 models require a 400 V 3 phase supply.

Specifications
  • Capacity:
    85 L
  • Description:
    Autoclave, top loading, 3870 ELV-WR-D, with water reservoir and advanced microprocessor control panel
  • Seq:
    009
  • Cat. No.:
    481-0905
  • Max. pressure:
    3,25 bar
  • W×D×H ext.:
    739×800×1075 mm
  • Temperature:
    134 °C
  • Model:
    3870 ELV-WR-D
  • Material:
    Stainless steel 316L
  • Int.Ø×H:
    380×690 mm
  • Power:
    3,3 kW
Services

Product Information

  • Catalog #
    Description
    For
    Availability
    Price
  • 481-0794
    Transportation from supplier to customer site, includes being unpacked and positioned at final destination with the end user (only available in Germany)
    50XX
    Log in for availability
    1 * 1 ST - SFr. 1’540.00

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Casters for 5075 autoclave
    Casters for 5075 autoclave
    TUTTADD222-0220
    Log in for availability
    1 * 1 ST - SFr. 420.00

Product Information

  • Catalog #
    For
    Availability
    Price
  • Replacement air filter for vertical autoclaves
    Replacement air filter for vertical autoclaves
    481-0855
    VHF-B
    Log in for availability
    1 * 1 ST - SFr. 337.00

Product Information

  • Catalog #
    For
    Availability
    Price
  • TUTTCLE096-0028
    All Tuttnauer models
    Log in for availability
    1 * 1 L - SFr. 27.40

Product Information

  • Catalog #
    For
    Availability
    Price
  • Printerpaper for thermal printer THE-002-0052 printer (5 rolls)
    Printerpaper for thermal printer THE-002-0052 printer (5 rolls)
    481-0851
    EL-ELV and D-line models
    Log in for availability
    1 * 5 ST - SFr. 13.90