ContactUsLinkComponent
An acetylated form of L-carnitine that facilitates the uptake of acetyl CoA into mitochondria during fatty acid oxidation, enhances acetylcholine production, and stimulates protein and membrane phospholipid synthesis 1 * 1 g
  CAYM16948-1
An acetylated form of L-carnitine that facilitates the uptake of acetyl CoA into mitochondria during fatty acid oxidation, enhances acetylcholine production, and stimulates protein and membrane phospholipid synthesis 1 * 1 g
  CAYM16948-1
Frequently Bought Together