Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Extraction thimbles, Munktell

Extraction thimbles, Munktell

26×80 mm
Codice catalogo BINZ400003
Nome del fornitore:   Ahlstrom
Extraction thimbles, Munktell
Codice catalogo BINZ400003
Nome del fornitore:  Ahlstrom
Extraction thimbles, Munktell
Extraction thimbles, Munktell

26×80 mm

Price
SFr. 209.00
Log in per disponibilità
1 * 25 ST
Product Details & Documents

Grade ET/401
Particularly suitable for the extraction of organic combinations out of reaction mixtures, food, natural substances, lacquer and binder analysis in paint colours, dust and tar determination in gases.
Max. Temperature: 120 °C.

Grade MK 360
A special development for the analysis of air pollution.
Max. Temperature: 900 °C

Grade MG160
Filter thimbles of high purity particularly recommended for aggressive solvents. For analytical registration of environmental pollution.
Max. Temperature: 500 °C

T* indicates is used for Tecator machines

Specifiche
  • Int.Ø×H:
    26×80 mm
  • Seq:
    31
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..