Order Entry
Switzerland
Orders LinkContactUsLinkComponent
β-Alanine

β-Alanine

Poly pail, 12 kg
Catalog # SPCMAL120-12KG
Supplier:   Spectrum Chemical
β-Alanine
Catalog # SPCMAL120-12KG
Supplier:  Spectrum Chemical
CAS Number:  107-95-9
β-Alanine

Poly pail, 12 kg

CAS Number:  107-95-9
Price
SFr. 2’370.00
Log in for availability
1 * 12 kg
Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg

Specifications