Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethyl 4-hydroxybenzoate NF

Ethyl 4-hydroxybenzoate NF

Fiber drum, 50 kg
Catalog # SPCMET117-50KGBL
Supplier:   Spectrum Chemical
Ethyl 4-hydroxybenzoate NF
Catalog # SPCMET117-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  120-47-8
Ethyl 4-hydroxybenzoate NF

Fiber drum, 50 kg

CAS Number:  120-47-8
Price
Inquire For Price
Log in for availability
1 * 50 kg
Product Details & Documents
Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Merck Index:13,03869
  • Boiling Pt:297 °C (1013 hPa)
  • Melting Pt:116…118 °C
  • Density:1.078 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:204-399-4
Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    50 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.