Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2-(2-Ethoxyethoxy)ethyl acetate

2-(2-Ethoxyethoxy)ethyl acetate

Steel pail, 20 L
Catalog # SPCME1023-20LTMT
Supplier:   Spectrum Chemical
2-(2-Ethoxyethoxy)ethyl acetate
Catalog # SPCME1023-20LTMT
Supplier:  Spectrum Chemical
CAS Number:  112-15-2
2-(2-Ethoxyethoxy)ethyl acetate

Steel pail, 20 L

CAS Number:  112-15-2
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Diethylene glycol monoethyl ether acetate
  • CAS Number:112-15-2
  • Formula:C₈H₁₆O₄
  • MDL Number:MFCD00041928
  • Molecular Weight:176.21 g/mol
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:–25 °C
  • Density:1.0114 g/cm³ (20 °C)
  • EINECS:203-940-1
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Steel pail
  • Pk:
    20 L

Specifications