Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2-Ethoxyethyl acetate

2-Ethoxyethyl acetate

Steel pail, 20 L
Catalog # SPCME1024-20LTMT
Flammable
Health
Irritant
Supplier:   Spectrum Chemical
2-Ethoxyethyl acetate
Catalog # SPCME1024-20LTMT
Supplier:  Spectrum Chemical
CAS Number:  111-15-9
2-Ethoxyethyl acetate

Steel pail, 20 L

CAS Number:  111-15-9
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Ethylglycol acetate
  • CAS Number:111-15-9
  • Formula:C₆H₁₂O₃
  • MDL Number:MFCD00009251
  • Molecular Weight:132.16 g/mol
  • Merck Index:13,03787
  • Boiling Pt:156 °C (1013 hPa)
  • Melting Pt:–62 °C
  • Density:0.976 g/cm³ (20 °C)
  • Flash Pt:54 °C
  • EINECS:203-839-2
  • UN:1172
  • ADR:3,III
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Steel pail
  • Pk:
    20 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.