Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic, powder USP

Bismuth(III) nitrate basic, powder USP

50 kg
Catalog # SPCMBI150-50KGBL
Oxidizing
Irritant
Supplier:   Spectrum Chemical
Bismuth(III) nitrate basic, powder USP
Catalog # SPCMBI150-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  1304-85-4
Bismuth(III) nitrate basic, powder USP

50 kg

CAS Number:  1304-85-4
Price
SFr. 6’720.00
Log in for availability
1 * 50 kg
Product Details & Documents

Form: Powder

Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Density:4.928 g/cm³ (20 °C)
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Specifications

Specification Test Results

  • Assay:
    79% Min.

Specifications

  • Pk:
    50 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.