Cholesterol NF
Fiber drum, 50 kg
Catalog # SPCMCH135-50KGBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
BSE/TSE statement (available upon request) Allergen statement (available upon request)
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
: LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
- CAS Number:57-88-5
- Formula:C₂₇H₄₆O
- MDL Number:MFCD00003646
- Molecular Weight:386.66 g/mol
- Merck Index:13,02221
- Boiling Pt:360 °C (1013 hPa)
- Melting Pt:147…150 °C
- Density:1.07 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-353-2
Specifications
Specification Test Results
- ASSAY (DRIED BASIS):95.0 - 102.0 %
- MELTING RANGE:147° - 150°C
- SPECIFIC ROTATION:-34° to -38°
- SOLUBILITY IN ALCOHOL:No deposit or turbidity forms
- ACIDITY:max 0.3 ml
- LOSS ON DRYING:max 0.3%
- RESIDUE ON IGNITION:max 0.1%
- β-CHOLESTANOL (5a-CHOLESTAN-3β-ol,DIHYDROCHOLESTEROL):max 0.6 %
- DESMOSTEROL:max 4%
- LATHOSTEROL:max 2%
- 24-DEHYDROLATHOSTEROL (5a-CHOLESTA-7,24 DIEN--3β-ol ):max 0.2 %
- 4-METHYLCHOLEST-5-EN-3β-OL:max 0.5 %
- TOTAL IMPURITIES INCLUDING RELATED STEROLS:max 5%
- ELEMENTAL IMPURITIES:AS REPORTED
- IDENTIFICATION (A):SPECTRUM MATCHES REFERENCE
- IDENTIFICATION (B):-34° to -38°
- IDENTIFICATION (C):147 - 150 °C
Specifications
- Pack type:Fiber drum
- Pk:50 kg



