ContactUsLinkComponent
TEBU BIO EGFP MRNA (5MEC, PSI) ENHANCED GREEN FLUORESCENT PROTEIN MRNALENGTH: 996 NUCLEOTIDES BUFFER: 10MM TRIS-HCL, PH7.5 1 * 100 µG
Catalog # XXXX040L-6101-100
Supplier:  Chemicals
TEBU BIO EGFP MRNA (5MEC, PSI) ENHANCED GREEN FLUORESCENT PROTEIN MRNALENGTH: 996 NUCLEOTIDES BUFFER: 10MM TRIS-HCL, PH7.5 1 * 100 µG
Catalog # XXXX040L-6101-100
Supplier:  Chemicals
Frequently Bought Together