RK 1028, Schwingwanne 500 × 300 × 200mm, 28Liter, G 1/2, Spitzen-leistung 2 × 600max. W/Per., Strom-aufnahme 1,4A, Aussenabmessun gen (L × B × H) 535 × 325 × 400mm 1 * 1 ST
Catalog # BAND322-CH
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together