ContactUsLinkComponent
SONOREX DIGITEC DT 514 BH, ULTRASONIC BATH, 230 V~ 50/60 HZ, ID 325X300X200 MM 18,7 L, S/S DIGITAL DISPLAY TIMER 1, 2, 3, 4, 5, 1 0, 15,30 MIN AND CONTINUOUS, ULTRASONIC PEAK OUTPUT 860 W, HF OUTPUT 215 WEFF., HEATER 20 - 80 °C (68-176°F) THERM 1 * 1 items
Catalog # BAND3216-CH
SONOREX DIGITEC DT 514 BH, ULTRASONIC BATH, 230 V~ 50/60 HZ, ID 325X300X200 MM 18,7 L, S/S DIGITAL DISPLAY TIMER 1, 2, 3, 4, 5, 1 0, 15,30 MIN AND CONTINUOUS, ULTRASONIC PEAK OUTPUT 860 W, HF OUTPUT 215 WEFF., HEATER 20 - 80 °C (68-176°F) THERM 1 * 1 items
Catalog # BAND3216-CH
Frequently Bought Together