ContactUsLinkComponent
SONOREX DIGITEC DT 103 H, ULTRASONIC BATH, 230 V~ 50/60 HZ, ID 240X140X150 MM 4,0 L, S/S DIGITAL DISPLAY TIMER 1, 2, 3, 4, 5, 10 , 15,30 MIN AND CONTINUOUS, ULTRASONIC PEAK OUTPUT 480 W, HF OUTPUT 80 WEFF., HEATER 20 - 80 °C (68-176°F) THERMOST 1 * 1 items
Catalog # BAND3201-GB
SONOREX DIGITEC DT 103 H, ULTRASONIC BATH, 230 V~ 50/60 HZ, ID 240X140X150 MM 4,0 L, S/S DIGITAL DISPLAY TIMER 1, 2, 3, 4, 5, 10 , 15,30 MIN AND CONTINUOUS, ULTRASONIC PEAK OUTPUT 480 W, HF OUTPUT 80 WEFF., HEATER 20 - 80 °C (68-176°F) THERMOST 1 * 1 items
Catalog # BAND3201-GB
Frequently Bought Together