ContactUsLinkComponent
SONOREX SUPER RK 1040, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID DIA. 540 MM, 500 MM HIGH, 38 L, S/S TIMER 1-15 MIN AND CONTINU OUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 W, HF-OUTPUT: 300 WEFF. 1 * 1 items
Catalog # BAND319-GB
SONOREX SUPER RK 1040, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID DIA. 540 MM, 500 MM HIGH, 38 L, S/S TIMER 1-15 MIN AND CONTINU OUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 W, HF-OUTPUT: 300 WEFF. 1 * 1 items
Catalog # BAND319-GB
Frequently Bought Together