ContactUsLinkComponent
SONOREX SUPER RK 100 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 240X140X100 MM 3 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATI NG 30-80°C (86-176°F), ULTRASONIC PEAK OUTPUT 320 W, HF-OUTPUT: 80 WEFF. 1 * 1 items
Catalog # BAND312-CH
SONOREX SUPER RK 100 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 240X140X100 MM 3 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATI NG 30-80°C (86-176°F), ULTRASONIC PEAK OUTPUT 320 W, HF-OUTPUT: 80 WEFF. 1 * 1 items
Catalog # BAND312-CH
Frequently Bought Together