SONOREX SUPER RK 102 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 240X140X100 MM 3 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATI NG 30-80°C (86-176°F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 480 W, HF-OUTPUT: 120 WEFF. 1 * 1 items
Catalog # BAND303-CH
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together