ContactUsLinkComponent
SONOREX LONGLIFE RK 1028 CH, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 500X300X300 MM 45 L, OSCILLATING TANK OF 2 MM S/S AISI 3 16 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80°C (86-176°F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 1 * 1 items
Catalog # BAND143-CH
SONOREX LONGLIFE RK 1028 CH, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 500X300X300 MM 45 L, OSCILLATING TANK OF 2 MM S/S AISI 3 16 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80°C (86-176°F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 1 * 1 items
Catalog # BAND143-CH
Frequently Bought Together