ContactUsLinkComponent
SONOREX TECHNIK RM 110 UH, 25 KHZ, ULTRASONIC BATH, 400 V 3N 50/60 HZ, ID 600X450X450 MM 121,5 L, WELDED TANK 2 MM S/S AISI 316 T I TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 1X1000 WEFF., WITH OVERF 1 * 1 items
Catalog # BAND8240-GB
SONOREX TECHNIK RM 110 UH, 25 KHZ, ULTRASONIC BATH, 400 V 3N 50/60 HZ, ID 600X450X450 MM 121,5 L, WELDED TANK 2 MM S/S AISI 316 T I TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 1X1000 WEFF., WITH OVERF 1 * 1 items
Catalog # BAND8240-GB
Frequently Bought Together