ContactUsLinkComponent
SONOREX SUPER RK 103 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 240X140X150 MM 4 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATI NG 30-80°C (86-176°F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 560 W, HF-OUTPUT: 140 WEFF. 1 * 1 items
Catalog # BAND326-CH
SONOREX SUPER RK 103 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 240X140X150 MM 4 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATI NG 30-80°C (86-176°F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 560 W, HF-OUTPUT: 140 WEFF. 1 * 1 items
Catalog # BAND326-CH
Frequently Bought Together