Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS

Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS

Multi-parameter meter, handheld, Multi 3510 IDS, set U (without electrode), with case, accessories and armor SM Pro
Catalog # 662-2139
Supplier:   WTW
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Catalog # 662-2139
Supplier:  WTW
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS

Multi-parameter meter, handheld, Multi 3510 IDS, set U (without electrode), with case, accessories and armor SM Pro

Price
SFr. 1’760.00
Log in for availability
1 * 1 ST
Product Details & Documents

This compact portable multi-parameter instrument enables measurements of pH, conductivity and dissolved oxygen with intelligent digital sensors (IDS). It has a universal measurement channel for different parameters. The robust, waterproof design makes it ideal for measurements in the field and in the laboratory.

  • Well laid out menus make operation safe and easy
  • MultiLab® Importer software enables data acquisition via Excel®
  • QSC function for optimum tracking of sensor quality
  • Calibration records and additional information are stored in the sensor
  • Wireless-ready for IDS wireless modules (for cable-free operation)
  • Available as sets in cases with sensors and accessories

Delivery information: Single meter (without electrodes) is supplied with case, MultiLab® Importer software, batteries, USB cable and short manual. The sets (except for set U) additionally include sensors.
Set U includes: Meter, case, instruction manual, stand, beaker, buffer 4 and 7, 3 mol/l KCl, conductivitiy standard, CD-ROM, driver software for USB, rechargeable batteries, USB cable, power supply, armoring SM Pro.
Please note: A wireless module is required for use with the wireless electrodes. This module is not included and needs to be ordered separately.

Documents

Specifications
  • Description:
    Multi-parameter meter, handheld, Multi 3510 IDS, set U (without electrode), with case, accessories and armor SM Pro
  • Seq:
    06
  • Connections:
    1 measurement input
  • Cat. No.:
    662-2139
  • Compliance:
    IP 67
  • Model:
    Multi 3510 IDS
  • Outputs:
    Mini USB-B
  • Data logging capacity:
    500 (manual); 4500 (automatic)
  • Power supply:
    4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • Display:
    Graphic LCD, backlit
Accessories

Product Information

  • Catalog #
    Length
    Availability
    Price
  • AS/IDS-1,5, connection cable for IDS sensors with waterproof plug head
    AS/IDS-1,5, connection cable for IDS sensors with waterproof plug head
    662-1733
    1,5 m
    Log in for availability
    1 * 1 ST - SFr. 163.00

Product Information

  • Catalog #
    Availability
    Price
  • Adapter cable, 1,5 m, for connecting a SenTix® combination electrode with S7 plug head to a MultiLine® instrument
    Adapter cable, 1,5 m, for connecting a SenTix® combination electrode with S7 plug head to a MultiLine® instrument
    662-1863
    Log in for availability
    1 * 1 ST - SFr. 411.00

Product Information

  • Catalog #
    Type
    Conductivity
    Availability
    Price
  • IDS conductivity cell, TetraCon® 925, 4-cell graphite
    IDS conductivity cell, TetraCon® 925, 4-cell graphite
    663-0152
    TetraCon® 925, 4-cell graphite
    10 µS/cm - 2000 mS/cm
    Log in for availability
    1 * 1 ST - SFr. 911.00

Product Information

  • Catalog #
    Availability
    Price
  • QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
    QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
    662-2132
    Log in for availability
    1 * 1 ST - SFr. 55.10

Product Information

  • Catalog #
    Availability
    Price
  • Large field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C
    Large field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C
    662-2134
    Log in for availability
    1 * 1 ST - SFr. 468.00

Product Information

  • Catalog #
    Availability
    Price
  • Case, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C
    Case, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C
    662-2133
    Log in for availability
    1 * 1 ST - SFr. 417.00

Product Information

  • Catalog #
    Availability
    Price
  • Protective housing SM PRO, rubber, for handheld WTW meters
    Protective housing SM PRO, rubber, for handheld WTW meters
    662-1278
    Log in for availability
    1 * 1 ST - SFr. 142.00

Product Information

  • Catalog #
    Availability
    Price
  • IDS wireless module for plug head sensors, IDS WLM-S
    IDS wireless module for plug head sensors, IDS WLM-S
    665-0423
    Log in for availability
    1 * 1 ST - SFr. 291.00

Product Information

  • Catalog #
    Availability
    Price
  • IDS wireless module for IDS meters, IDS WLM-M
    IDS wireless module for IDS meters, IDS WLM-M
    665-0424
    Log in for availability
    1 * 1 ST - SFr. 234.00

Product Information

  • Catalog #
    Availability
    Price
  • USB charger for wireless modules
    USB charger for wireless modules
    665-0425
    Log in for availability
    1 * 1 ST - SFr. 197.00

Product Information

  • Catalog #
    Availability
    Price
  • Kit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supply
    Kit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supply
    662-2050
    Log in for availability
    1 * 1 ST - SFr. 639.00

Product Information

  • Catalog #
    Availability
    Price
  • Universal USB power supply
    Universal USB power supply
    662-2148
    Log in for availability
    1 * 1 ST - SFr. 55.30

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.