Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Product Details & Documents
This compact portable multi-parameter instrument enables measurements of pH, conductivity and dissolved oxygen with intelligent digital sensors (IDS). It has a universal measurement channel for different parameters. The robust, waterproof design makes it ideal for measurements in the field and in the laboratory.
- Well laid out menus make operation safe and easy
- MultiLab® Importer software enables data acquisition via Excel®
- QSC function for optimum tracking of sensor quality
- Calibration records and additional information are stored in the sensor
- Wireless-ready for IDS wireless modules (for cable-free operation)
- Available as sets in cases with sensors and accessories
Delivery information: Single meter (without electrodes) is supplied with case, MultiLab® Importer software, batteries, USB cable and short manual. The sets (except for set U) additionally include sensors.
Set U includes: Meter, case, instruction manual, stand, beaker, buffer 4 and 7, 3 mol/l KCl, conductivitiy standard, CD-ROM, driver software for USB, rechargeable batteries, USB cable, power supply, armoring SM Pro.
Please note: A wireless module is required for use with the wireless electrodes. This module is not included and needs to be ordered separately.
Documents
MultiLine® IDS Portable Meters
Product Literature
Specifications
- Description:Multi-parameter meter, handheld, Multi 3510 IDS, set 4, with FDO® 925 (optical DO sensor)
- Seq:05
- Connections:1 measurement input
- Cat. No.:662-2138
- Compliance:IP 67
- Model:Multi 3510 IDS
- Outputs:Mini USB-B
- Data logging capacity:500 (manual); 4500 (automatic)
- Power supply:4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
- Display:Graphic LCD, backlit
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- AS/IDS-1,5, connection cable for IDS sensors with waterproof plug headAS/IDS-1,5, connection cable for IDS sensors with waterproof plug head662-17331,5 mLog in for availability1 * 1 ST - SFr. 163.00
Product Information
- Catalog #AvailabilityPrice
- Adapter cable, 1,5 m, for connecting a SenTix® combination electrode with S7 plug head to a MultiLine® instrumentAdapter cable, 1,5 m, for connecting a SenTix® combination electrode with S7 plug head to a MultiLine® instrument662-1863Log in for availability1 * 1 ST - SFr. 411.00
Product Information
- Catalog #TypeConductivityAvailabilityPrice
- IDS conductivity cell, TetraCon® 925, 4-cell graphiteIDS conductivity cell, TetraCon® 925, 4-cell graphite663-0152TetraCon® 925, 4-cell graphite10 µS/cm - 2000 mS/cmLog in for availability1 * 1 ST - SFr. 911.00
Product Information
- Catalog #AvailabilityPrice
- QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)662-2132Log in for availability1 * 1 ST - SFr. 55.10
Product Information
- Catalog #AvailabilityPrice
- Large field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °CLarge field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C662-2134Log in for availability1 * 1 ST - SFr. 468.00
Product Information
- Catalog #AvailabilityPrice
- Case, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °CCase, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C662-2133Log in for availability1 * 1 ST - SFr. 417.00
Product Information
- Catalog #AvailabilityPrice
- Protective housing SM PRO, rubber, for handheld WTW metersProtective housing SM PRO, rubber, for handheld WTW meters662-1278Log in for availability1 * 1 ST - SFr. 142.00
Product Information
- Catalog #AvailabilityPrice
- IDS wireless module for plug head sensors, IDS WLM-SIDS wireless module for plug head sensors, IDS WLM-S665-0423Log in for availability1 * 1 ST - SFr. 291.00
Product Information
- Catalog #AvailabilityPrice
- IDS wireless module for IDS meters, IDS WLM-MIDS wireless module for IDS meters, IDS WLM-M665-0424Log in for availability1 * 1 ST - SFr. 234.00
Product Information
- Catalog #AvailabilityPrice
- USB charger for wireless modules
Product Information
- Catalog #AvailabilityPrice
- Kit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supplyKit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supply662-2050Log in for availability1 * 1 ST - SFr. 639.00
Product Information
- Catalog #AvailabilityPrice
- Universal USB power supply
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



