Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
This compact portable multi-parameter instrument enables measurements of pH, conductivity and dissolved oxygen with intelligent digital sensors (IDS). It has a universal measurement channel for different parameters. The robust, waterproof design makes it ideal for measurements in the field and in the laboratory.
- Well laid out menus make operation safe and easy
- MultiLab® Importer software enables data acquisition via Excel®
- QSC function for optimum tracking of sensor quality
- Calibration records and additional information are stored in the sensor
- Wireless-ready for IDS wireless modules (for cable-free operation)
- Available as sets in cases with sensors and accessories
: Single meter (without electrodes) is supplied with case, MultiLab® Importer software, batteries, USB cable and short manual. The sets (except for set U) additionally include sensors.
Set U includes: Meter, case, instruction manual, stand, beaker, buffer 4 and 7, 3 mol/l KCl, conductivitiy standard, CD-ROM, driver software for USB, rechargeable batteries, USB cable, power supply, armoring SM Pro.
Please note: A wireless module is required for use with the wireless electrodes. This module is not included and needs to be ordered separately.
- Description:Multi-parameter meter, handheld, Multi 3510 IDS, set 3, with TetraCon® 925 (4-electrode conductivity cell)
- Seq:04
- Connections:1 measurement input
- Cat. No.:662-2137
- Compliance:IP 67
- Model:Multi 3510 IDS
- Outputs:Mini USB-B
- Data logging capacity:500 (manual); 4500 (automatic)
- Power supply:4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
- Display:Graphic LCD, backlit
Accessories
- DescriptionLength
- 662-1733AS/IDS-1,5, connection cable for IDS sensors with waterproof plug head1,5 m1 * 1 ST - SFr. 163.00
- Description
- 662-1863Adapter cable, 1,5 m, for connecting a SenTix® combination electrode with S7 plug head to a MultiLine® instrument1 * 1 ST - SFr. 411.00
- DescriptionTypeConductivity
- 663-0152IDS conductivity cell, TetraCon® 925, 4-cell graphiteTetraCon® 925, 4-cell graphite10 µS/cm - 2000 mS/cm1 * 1 ST - SFr. 911.00
- Description
- 662-2132QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)1 * 1 ST - SFr. 55.10
- Description
- 662-2134Large field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C1 * 1 ST - SFr. 468.00
- Description
- 662-2133Case, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C1 * 1 ST - SFr. 417.00
- Description
- Description
- Description
- Description
- Description
- 662-2050Kit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supply1 * 1 ST - SFr. 639.00
- Description
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



