ContactUsLinkComponent
Ferritin forms a 24-subunit cage for storage of mineral iron. In plants, ferritin is predominantly localized in the plastids, and it s expression is upregulated in response to iron or oxidative stress. 1 * 50 µG
Catalog # AGRIAS152898
Supplier:  Agrisera
undefined
Ferritin forms a 24-subunit cage for storage of mineral iron. In plants, ferritin is predominantly localized in the plastids, and it s expression is upregulated in response to iron or oxidative stress. 1 * 50 µG
Catalog # AGRIAS152898
Supplier:  Agrisera