ContactUsLinkComponent
Temperature Module TM-LI-C48 for HAAKE Viscotester iQ *********************************************** Universal plug-and-play temperature for reliable and accurate temperature control using a circulator. Temperature range*: Coaxial cylinders: -20 C 1 * 1 items
Catalog # HAAK222-2173
Supplier:  HAAKE
Temperature Module TM-LI-C48 for HAAKE Viscotester iQ *********************************************** Universal plug-and-play temperature for reliable and accurate temperature control using a circulator. Temperature range*: Coaxial cylinders: -20 C 1 * 1 items
Catalog # HAAK222-2173
Supplier:  HAAKE
Frequently Bought Together