REGIS COLUM IAM DD2 10CM X 4.6MM ID 1 * 10 µG
Catalog # HISM1-774011-300
Supplier: HICHROM
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Pipette stand, linear
Supplier: Sartorius
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection