ACID NUMBER STANDARD 0.1 MGKOH/G. REFERENCE MATERIALS INTENDED FOR USE IN THE DETERMINATION OF ACID 1 * 800 g
Catalog # PROOVHGAN-0.1-800G
Supplier: LGC Standards PROMOCHEM
ACID NUMBER STANDARD 0.1 MGKOH/G. REFERENCE MATERIALS INTENDED FOR USE IN THE DETERMINATION OF ACID 1 * 800 g
Catalog # PROOVHGAN-0.1-800G
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck