ContactUsLinkComponent
COMPREHENSIVE MIX A: AG, AL, AS, BA, BI, CA, CD, CE, DY, ER, EU, GA, GD, HO, LA, LU, MG, NA, ND, P, PB, PR, RB, SC, SE, SM, SR, TB, TH, TL, TM, U, Y, YB @ 10µG/ML IN 40% AQUA REGIA 1 * 100 mL
Catalog # PROOVHG-SM80C-100
COMPREHENSIVE MIX A: AG, AL, AS, BA, BI, CA, CD, CE, DY, ER, EU, GA, GD, HO, LA, LU, MG, NA, ND, P, PB, PR, RB, SC, SE, SM, SR, TB, TH, TL, TM, U, Y, YB @ 10µG/ML IN 40% AQUA REGIA 1 * 100 mL
Catalog # PROOVHG-SM80C-100
Frequently Bought Together