ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, A, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300151A011
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ29/32, A, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300151A011
Supplier:  BUCHI
Frequently Bought Together