ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, A, I-300P, 230V. . Lift System: Elec. Heating Bath: w/o SJ 29/32 Glass set: Diagonal (A) int.: I- 300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300201A031
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, A, I-300P, 230V. . Lift System: Elec. Heating Bath: w/o SJ 29/32 Glass set: Diagonal (A) int.: I- 300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300201A031
Supplier:  BUCHI
Frequently Bought Together