ContactUsLinkComponent
Rotavapor R-300 no bath, no joint., V, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: Vertical (V) int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300200V051
Supplier:  BUCHI
Rotavapor R-300 no bath, no joint., V, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: Vertical (V) int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300200V051
Supplier:  BUCHI
Frequently Bought Together