ContactUsLinkComponent
Rotavapor R-300 no bath, no joint., no glass, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: w/o int.: I-300 Pr o, valve unit 1 * 1 items
Catalog # BUCH11R300200N051
Supplier:  BUCHI
Rotavapor R-300 no bath, no joint., no glass, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: w/o int.: I-300 Pr o, valve unit 1 * 1 items
Catalog # BUCH11R300200N051
Supplier:  BUCHI
Frequently Bought Together