ContactUsLinkComponent
Rotavapor R-300 w safety coated vert cond ass, Elec. jack. Bath B-305 with a dig display and temp range to 220°C. Max flask size 500 0mL. Evap flask 1L. Rec flask 1L.SJ 29/32. Inc. V-300 vac pump and I-300 Pro int. 1 * 1 items
Catalog # BUCH11SR300251VP01
Supplier:  BUCHI
Rotavapor R-300 w safety coated vert cond ass, Elec. jack. Bath B-305 with a dig display and temp range to 220°C. Max flask size 500 0mL. Evap flask 1L. Rec flask 1L.SJ 29/32. Inc. V-300 vac pump and I-300 Pro int. 1 * 1 items
Catalog # BUCH11SR300251VP01
Supplier:  BUCHI
Frequently Bought Together