ContactUsLinkComponent
Autoclave for sterilisation at 125°C or 140°C. Venting by stream process. Manual process timing with integrated timer. Process control by thermostat and precision valve. 18 liter volume. 1 * 1 items
Catalog # CERT8510174_B
Supplier:  CERTOCLAV
Autoclave for sterilisation at 125°C or 140°C. Venting by stream process. Manual process timing with integrated timer. Process control by thermostat and precision valve. 18 liter volume. 1 * 1 items
Catalog # CERT8510174_B
Supplier:  CERTOCLAV
Frequently Bought Together