ContactUsLinkComponent
Software conforming to FDA “AtmoCONTROL“. Meets the requirements for the use of electronically stored data sets and electronic signa tures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence for the co 1 * 1 items
Catalog # MEMMFDAQ1UF750TS
Supplier:  MEMMERT
Software conforming to FDA “AtmoCONTROL“. Meets the requirements for the use of electronically stored data sets and electronic signa tures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence for the co 1 * 1 items
Catalog # MEMMFDAQ1UF750TS
Supplier:  MEMMERT
Frequently Bought Together