Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Propionic acid ACS

Propionic acid ACS

Glass bottle, 500 mL
Catalog # ACRO447235000
Corrosive
Flammable
Irritant
Propionic acid ACS
Catalog # ACRO447235000
CAS Number:  79-09-4
Propionic acid ACS
Propionic acid ACS

Glass bottle, 500 mL

CAS Number:  79-09-4
Price
SFr. 231.00
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:141 °C (760 mmHg)
  • Melting Pt:–22 °C
  • Density:0.99 g/cm³
  • Flash Pt:51 °C
  • Storage Temperature:Ambient
  • UN:3463
  • ADR:8,II

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.15 %
  • Color scale:
    ≤20 APHA
  • Residue after evaporation:
    ≤0.01 %
  • Titration with NaOH:
    ≥99.5 %
  • Heavy metals (as Pb):
    ≤0.001 %
  • Carbonyl compounds:
    ≤0.002 %
  • Oxidisable substance (HCOOH):
    ≤0.1 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.