Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Propionic acid ACS | Avantor

Propionic acid ACS

Glass bottle, 100 mL
Katalognr. ACRO447231000
Ätzend
Entzündlich
Reizend
Propionic acid ACS
Katalognr. ACRO447231000
CAS Nummer:  79-09-4
Propionic acid ACS
Propionic acid ACS

Glass bottle, 100 mL

CAS Nummer:  79-09-4
Preis
SFr. 66.60
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents
Synonyme:  Propanoic acid
  • CAS Nummer:79-09-4
  • Formel:C₃H₆O₂
  • MDL-Nummer:MFCD00002756
  • Molekulargewicht:74.08 g/mol
  • Siedepunkt:141 °C (760 mmHg)
  • Schmelzender Platingehalt:–22 °C
  • Dichte:0.99 g/cm³
  • Blitz Pt:51 °C
  • Lagertemperatur:Ambient
  • UN:3463
  • ADR:8,II

Dokumente

Zertifikate
Suche nach Chargennummer
Technische Daten

Ergebnisse des Spezifikationstests

  • Appearance:
    Clear liquid
  • Water:
    ≤0.15 %
  • Color scale:
    ≤20 APHA
  • Residue after evaporation:
    ≤0.01 %
  • Titration with NaOH:
    ≥99.5 %
  • Heavy metals (as Pb):
    ≤0.001 %
  • Carbonyl compounds:
    ≤0.002 %
  • Oxidisable substance (HCOOH):
    ≤0.1 %

Technische Daten

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.