Vanadium(V) oxide 99+%
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Vanadium(V) oxide 99+%
Catalog # ACRO450162500
Supplier: Thermo Fisher Scientific
CAS Number: 1314-62-1
Jump to:
Product Details & Documents
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Boiling Pt:1750 °C (760 mmHg)
- Melting Pt:690 °C
- Density:3.35 g/cm³
- Storage Temperature:Ambient
- UN:2862
- ADR:6.1,III
Documents
ACRO45016
SDS
Specifications
Specification Test Results
- Appearance:Dark yellow Powder
- Assay:≥99.0 %
- Particle size:≤1.2 mm
- Chromium (Cr):≤0.1 %
- Iron (Fe):≤0.1 %
- Phosphorus (P):≤0.04 %
- Potassium (K):≤0.4 %
- Silicon (Si):≤0.06 %
- Sodium (Na):≤0.3 %
- Sulfur (S):≤0.07 %
Specifications
- Pack type:Plastic bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



