Order Entry
Switzerland
Orders LinkContactUsLinkComponent
p-Xylene 99% for HPLC

p-Xylene 99% for HPLC

Glass bottle, 100 mL
Catalog # ACRO445681000
Flammable
Irritant
Health
p-Xylene 99% for HPLC
Catalog # ACRO445681000
CAS Number:  106-42-3
p-Xylene 99% for HPLC
p-Xylene 99% for HPLC

Glass bottle, 100 mL

CAS Number:  106-42-3
Price
SFr. 33.50
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  1,4-Dimethylbenzene
  • CAS Number:106-42-3
  • Formula:C₆H₄(CH₃)₂
  • MDL Number:MFCD00008556
  • Molecular Weight:106.17 g/mol
  • Merck Index:12,10214
  • Boiling Pt:138 °C (760 mmHg)
  • Melting Pt:13 °C
  • Density:0.866 g/cm³
  • Flash Pt:25 °C
  • Storage Temperature:Ambient
  • EINECS:203-396-5
  • UN:1307
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 %
  • GC:
    ≥98.5 %
  • UV:
    at 294 nm A: ≤1.0
  • Color scale:
    ≤15 APHA
  • Residue after evaporation:
    ≤0.0005 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.